Key Documents
Z683450
BRAND® seripettor® dispensing cartridges
capacity 10 mL, sterile
About This Item
UNSPSC Code:
41121500
NACRES:
NB.43
material
polyethylene piston
polypropylene cylinder
sterility
sterile
packaging
pkg of 7 ea individually wrapped
manufacturer/tradename
BRAND
capacity
10 mL
Other Notes
BR704506 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG