Key Documents
Z682829
Ace receiving flask,
capacity 1000 mL, joint: ST/NS 35/20, glass
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
glass
round bottom flask
single-neck
Joint
joint: ST/NS35/20
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Büchi equivalent, 00425
capacity
1000 mL
General description
The receiving flasks have the standard joint to fit the glassware used in rotary evaporators, and are available with poly-coating or plain glass.