Key Documents
Z682314
Ace rotary evaporator condenser complete "A" assembly
plastic-coated glass, fits Büchi® models R-200/R-205 and R-114/R-144
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
plastic-coated glass
manufacturer/tradename
Büchi equivalent, 40633/32304
Related Categories
General description
The complete "A" evaporator condenser assembly includes the main glass condenser body with inner coil for angled installation, GL threaded caps and hose barbs. Bottom joint is 35/20. It is designed to fit Büchi® evaporator models 200/205 and series 114-144.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)