Key Documents
Z682209
Ace receiving flask adapter
joint: ST/NS 29/32, fits Büchi® models R-200/R-205
Synonym(s):
Ace receiving flask adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Joint
joint: ST/NS29/32
manufacturer/tradename
Büchi equivalent, 40615
General description
The receiving flask adapter fits condenser assemblies "C", "CR", "A", and "V" for Büchi® evaporator models R-200/205. Adapter clip: not included.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG