Key Documents
Z679127
Anti-static unisex laboratory coats
size M
Synonym(s):
laboratory apparel
About This Item
UNSPSC Code:
46181532
NACRES:
NB.35
size
M
feature
CE compliant
availability
not available in USA and Canada
General description
Tough white 210 g/m cotton/polyester reusable laboratory coats, full-length with a Nega-stat grid offering electrostatic protection.
  • Concealed stud-fastening front
  • Two hip pockets and one breast pocket, all reinforced with bar tacks
  • Finished length 100 cm, with center back vent
Conforms to prEN1149-3:2001