Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z678465

Krytox® vacuum oil

grade, 1525

Synonym(s):

Krytox Lubricant, Vacuum Oil

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161501
NACRES:
NB.79

mol wt

3470

feature

grade 1525

manufacturer/tradename

Du Pont

viscosity number

(20 °C) (261 mm2/s)

density

1.90 g/mL at 20 °C

Looking for similar products? Visit Product Comparison Guide

General description

DuPont Krytox vacuum pump fluids are used in applications where conventional vacuum pump oils cause safety, waste disposal and maintenance problems. They are nonflammable and eliminate the chance of fire in pumps. They are nonreactive and safe to use in oxygen systems. This gives Krytox superior stability as a vacuum pump fluid.

Krytox vacuum fluids are precisely distilled to provide low vapor pressures and give superior performance.
  • Nonflammable
  • Nonreactive
  • Free from acetal groups
  • Recyclable

Legal Information

Krytox is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service