Key Documents
Z678139
Finnpipette® F3 variable volume pipette
volume 20-200 μL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
color-coded yellow
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Finn 4640050
volume
20-200 μL
General description
Finnpipette F3 has a double action pipetting button with rotating upper part enabling smooth and steady pipetting action at the same time as preventing accidental volume alterations. Finpipette F3 is equipped with the Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Double action pipetting button with very light and smooth pipetting action
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Color coded for easy tip volume recognition
Compatibility
Uses Finntip 250 Universal or Flex 300
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)