Key Documents
Z677612
Camatril nitrile long cuff gloves
size XL
Synonym(s):
disposable glove
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
Pricing and availability is not currently available.
material
green gloves
green nitrile
size
XL
sterility
non-sterile
feature
CE compliant
disposable
latex free
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
packaging
pack of 1 pair
manufacturer/tradename
KCL 733
KCL 733
L × thickness
600 mm × 0.5 mm
General description
Camatril nitrile long cuff gloves are of high standard, with a wide range of applications and made of specific high quality Nitrile. The large spectrum of chemical resistance ensures the use of these gloves in numerous fields. The remarkably high abrasion, cut and puncture resistance allows work with sharp edged materials. Good flexibility, anatomical design and excellent fit ensure a good sense of touch.
  • Ergonomic fit and high elasticity ensures good sensitivity
  • Roughened palm for firm grip when wet
  • High mechanical resistance
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • CE compliant
Legal Information
Camatril is a trademark of KCL GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)