Key Documents
Z615544
Julabo® bath covers
Lift-up Makrolon cover to fit TW20
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970288
L × W × H
35 cm × 51 cm × 16 cm
General description
Lift-up Makrolon batch cover W 51cm x L35 cm x H 16cm, use to preventnt liquid losses due to evaporation, protects samples from contamination. For use with Julabo eater bath models SW22, SW23.
Features and Benefits
  • can be used up to +80°C
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)