Documents
Z567957
J-KEM® water flow monitor
AC/DC input 230 V AC
About This Item
UNSPSC Code:
20121904
NACRES:
NB.51
description
requires a flow sensor
output
2300 watts
General description
  • Digital sensor measures actual coolant flow rate
  • Optional audible alarm
Precisely monitors the flow rate of water through a condenser, water bath, or other piece of equipment. If the flow rate falls below the minimum rate set by the user (i.e. loss of pressure, broken hose), power is turned off to the monitor′s output receptacle and any equipment plugged into it. Power remains off until being manually reset. The optional water shut off valve stops water flow during an alarm condition preventing a flooded lab. With safety as its primary goal, the J-KEM Water Flow Monitor measures the actual water flow rate, rather than back pressure as in commonly measured by lower quality instruments.
Legal Information
J-KEM is a registered trademark of J-KEM Electronics, Inc.