Key Documents
Z562513
LabJaws jumbo clamp holder
jaw capacity 0-21 mm , 0-83 inches, aluminum
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
material
aluminum
manufacturer/tradename
Troemner 916050
jaw capacity
0-21 mm , 0-83 inches
General description
Corrosion resistant. Has precise fit and powerful holding ability for heavy-duty applications. Ideal for holding clamps to lab frames or ring stands.
Legal Information
LabJaws is a trademark of Henry Troemner LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)