Key Documents
Z552909
Sigma-Aldrich
Aldrich® reaction vessel bubbler with SafetyBarb® hose connectors
joint: ST/NS 29/32
Synonym(s):
bubblers, scientific bubblers
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS29/32
capacity
5-7 mL
General description
Excellent from closing off reaction vessel from the atmosphere. Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)