Key Documents
Z551821
Sigma-Aldrich
Aldrich® Thermometer Adapter
offset, male joint: ST/NS 24/40, GL 14 thread
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Pricing and availability is not currently available.
Joint
male joint: ST/NS24/40
fitting
GL 14 thread
General description
Has PTFE faced silicone washer and open-top cap.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)