Key Documents
Z543209
Sigma-Aldrich
Periodic table of the elements
Poster size
About This Item
UNSPSC Code:
60104506
NACRES:
CE.34
W × L
24 in. × 36 in.
General description
Laminated. Excellent reference tools for students and researchers.
Poster:
Full-color. Chart information includes: atomic weight, melting point, boiling point, density, electronegativity, first ionization potential, group classifications, atomic number, oxidation states, element symbol, electronic configuration, and name. Plus special tables for Elementary Subatomic Particles and % Ionic Character of a Single Chemical Bond.
Binder:
Two-sided chart features more than 20 properties of the elements, from electron configuration and electronegativity to covalent radius and thermal conductivity.
Wallet:
Two-sided chart features properties of the elements in table format with useful chemical constants on the back.
Choose from one of the most recent versions:
Certificates of Analysis (COA)