Key Documents
Z529656
Sigma-Aldrich
Digital Wig-L-Bug® grinder/mixer
low profile
About This Item
UNSPSC Code:
12000000
AC/DC input
110 V
availability
not available in EU
W × D × H
11 1/2 in. × 8 in. × 4 1/2 in.
General description
  • Easy-to-use icon-driven control panel
  • Membrane keypad is easily cleaned and disinfected
  • Safety cover shields capsules, reduces noise
  • Speeds of 3,800, 4,200, 4,600, and 4,800 rpm
  • Accurate digital circuitry with LED readout of time and speed
  • Vials and pestles sold separately
Legal Information
Wig-L-Bug is a registered trademark of Dentsply Rinn
Choose from one of the most recent versions:
Certificates of Analysis (COA)