Key Documents
Z510807
Sigma-Aldrich
Connecting Adapter
female joint: ST/NS 24/40, thread size 13-425
Synonym(s):
747130-1324
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
thread size
13-425
Joint
female joint: ST/NS24/40
General description
Connects ground joint glassware to threaded microscale component or sample vials. Excellent for use with splash-guards to recover small quantities of material during rotary evaporation. Order threaded vial connectors separately.
Features and Benefits
• Accessory for microscale MICROFLEX threaded kits• Connects Standard Taper outer ground joint to GPI thread of inner ground joint• Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements