Key Documents
Z414255
Sigma-Aldrich
Aldrich® septum-inlet adapter, in-line
Plain, Joints: ST/NS 24/40 (2)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
Joints: ST/NS24/40 (2)
Related Categories
General description
Inlet diam. 14 mm. Requires septum Z564702 or Z564699.
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC