Documents
Z402362
Sigma-Aldrich
Best® butyl gloves
size XL
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
black gloves
butyl rubber
size
XL
sterility
non-sterile
feature
latex free
powder-free
reusable
packaging
pkg of 1 pair
manufacturer/tradename
Best Manufacturing 878-10
L × thickness
350 mm × 0.70 mm
Features and Benefits
  • Impermeable
  • Resistant to water
  • Great tactility and performance
  • Provides permeation resistance to gases and water vapors
  • Unsupported, heavy-duty, 100% butyl rubber
  • Injection molded
  • Smooth grip with rolled cuff
  • Offers flexibility and durability
  • Excellent resistance to ketones, acids, caustics, isocyanate, and gases
Legal Information
Best is a registered trademark of Showa Glove Co.