Key Documents
Z285501
Sigma-Aldrich
DigiTrol II temperature controller
AC/DC input 240 V AC, 10 A (CE Compliant)
About This Item
UNSPSC Code:
41112303
NACRES:
NB.76
manufacturer/tradename
Glas-Col 104APL624K (CE)
parameter
32-999 °F temperature (0-750 °C)
output
2,400 watts
General description
Simultaneously displays set point and bath temperature to 1.0°C. Supplied with a Type K, 6 in. immersion thermocouple probe that fits Aldrich oil baths. 240 V models are supplied with a US connector.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)