Documents
Z280364
Sigma-Aldrich
PVC laboratory tubing
I.D. × O.D. 3/16 in. × 5/16 in.
Synonym(s):
Saint Gobain Versilon ABH02012
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
packaging
pkg of 100 feet
I.D. × O.D.
3/16 in. × 5/16 in.
wall size
1/16 in.
General description
General-purpose polyvinyl chloride laboratory tubing. Clear, flexible tubing ideal for drains and rinse tubes. Max. temp.: 165°F (73.4°C).