Key Documents
Z275301
Sigma-Aldrich
Cannon-Fenske viscometer tube
size 100
Synonym(s):
Viscosity Measurement Device, Viscosity Tube
About This Item
UNSPSC Code:
41111700
NACRES:
PB.36
size
100
description
constant ~ 0.015 cSt/s
manufacturer/tradename
Ace Glass 798839
viscosity measuring range
3-15 cSt
Related Categories
General description
Obtain kinematic viscosities using tubes designed for ASTM Method D445 and D2515. Viscosities can be measured with a precision of ±0.2% quickly and easily.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.
Choose from one of the most recent versions:
Certificates of Analysis (COA)