Key Documents
Z261319
Stainless steel 316 syringe needle, noncoring point
gauge 18, L 6 in.
About This Item
UNSPSC Code:
42142600
eCl@ss:
32070500
NACRES:
NB.03
description
Micro-Mate Hub with 316 Stainless Steel Tubing
packaging
pack of 1 ea
manufacturer/tradename
Popper 9860
L
6 in.
gauge
18
General description
Specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. Female Luer hubs and needles are made of 316 SS. Noncontaminating, easy to clean and resistant to corrosion.
Micro-Mate Hub with 316 Stainless Steel Tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)