Key Documents
Z260266
FORTUNA® Universal Optifix® HF dispenser
HF model, volume 2-10 mL
Synonym(s):
HF Dispenser
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
subdivision 0.2 mL
manufacturer/tradename
Fortuna Graf 101.106-38
volume measuring range
2-10 mL
volume
2-10 mL
fitting
GL 40
GL 45
General description
For all liquids that can be handled with the Optifix dispensers and for hydrofluoric acid (HF)
Features:
  • Dispensing cylinder made of PTFE
  • Massive PTFE encased dispensing piston
  • FIX adapter made of PTFE
  • Ergonomic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
  • Valve block made of PTFE, with additional safety valve in the ventilation tube, no metal springs in the valves - no corrosion
Supplied:
FORTUNA Universal Optifix HF dispenser, with 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
Legal Information
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)