Key Documents
Z260223
Sigma-Aldrich
FORTUNA® Optifix® Safety bottle-top dispenser
Safety model, volume 6-30 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
subdivision 0.5 mL
manufacturer/tradename
Fortuna Graf 101.107-44
volume measuring range
6-30 mL
volume
6-30 mL
fitting
GL 40
GL 45
General description
For dispensing strong acids and aggressive chemicals
Features:
  • PTFE encased dispensing piston with sealing lip at its end
  • FIX adapter made of PTFE
  • Valve block made of PTFE, no metal springs in the valve - no corrosion
  • Visible dispensing process by the use of a dispensing cylinder made of glass, coated with plastic-protection against breakage
  • Ergonmic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
Supplied as Optifix® Safety dispenser, 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
Legal Information
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)