Key Documents
Z249602
Sigma-Aldrich
Aldrich® modified, jacketed Dean-Stark trap
joint: ST/NS 24/40, Overall H 300 mm
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Joint
joint: ST/NS24/40
Overall H
300 mm
General description
Designed specifically for azeotropic distillation using solvents heavier than water (e.g., chloroform). 350 mL capacity. 0-3 mm PTFE valve.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC