Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z243906

Supelco

Aldrich® TLC developing tank - cylindrical

glass tank, O.D. × H 6.5 cm × 10.5 cm

Synonym(s):

TM=["Aldrich"] Cylindrical Tank

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NC.07

technique(s)

thin layer chromatography (TLC): suitable

O.D. × H

6.5 cm × 10.5 cm

Looking for similar products? Visit Product Comparison Guide

General description

Can be used as a staining chamber. Made of glass with lid. Compact size offers good plate visibility and minimal solvent use.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

replacement part

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service