Key Documents
Z230855
Sigma-Aldrich
Aldrich® Schlenk-type male joint cap
male joint: ST/NS 14/20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
label
L
Joint
male joint: ST/NS14/20
Related Categories
General description
Thermoplastic screw caps with Viton O-rings usable to 140°C. This stopper has a rounded top and fits where clearances are tight.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.