Documents
Z225339
Sigma-Aldrich
Aldrich® SAFE-PURGE valve
automatic
About This Item
UNSPSC Code:
40141600
NACRES:
NB.79
parameter
>5 psi pressure
General description
Used with purge gas pressure greater than 5 psi. Should any oil pass the check valve, the spring-loaded plug will prevent any oil or air from entering the system.
The Aldrich SAFE-PURGE valves provide for safe, high-performance operation. Both the manual and automatic spring-loaded styles feature a durable PTFE metering valve.
  • Quick and efficient purging of reaction vessel which conserves expensive purge gases
  • Safely vents toxic gases from built-in oil bubbler
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SAFE-PURGE is a trademark of Sigma-Aldrich Co. LLC