Key Documents
Z218901
Ace giant extraction thimble
I.D. × L 68 mm × 250 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
manufacturer/tradename
Ace Glass 681204
I.D. × L
68 mm × 250 mm
Related Categories
General description
For use with Ace giant Soxhlet extractors. Thimbles are readily permeable to the flow of ether and other organic solvents. Made of cellulose.