Key Documents
Z203378
Sigma-Aldrich
Aldrich® vacuum-filtration adapter
male joint: ST/NS 29/32, flange I.D. 38 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
male joint: ST/NS29/32
flange I.D.
38 mm
Related Categories
General description
Enables filtration (via a Buchner funnel) directly into a single-neck, round-bottom flask. Includes rubber sleeve.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
replacement part
Choose from one of the most recent versions:
Certificates of Analysis (COA)