Key Documents
Z158038
Sigma-Aldrich
Spilfyter® hazmat spill control, polypropylene
Pillow, capacity 250 mL
About This Item
UNSPSC Code:
47131905
NACRES:
NB.35
packaging
pkg of 20 ea
W × L
10 in. × 10 in.
capacity
250 mL
General description
Spilfyter® products are made from a specially formulated, lint-free, 100% PP fabric that is virtually inert and will absorb any liquid with which they come in contact. They are able to collect up to 10 times their own weight in waste material (depending upon its viscosity) in 30s. Saturated Spilfyter® products can be landfilled, neutralized, or incinerated in accordance with federal, state and local regulations.
Legal Information
Spilfyter is a registered trademark of National Packaging Services Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)