Key Documents
Z156523
Sigma-Aldrich
Pinch clamps for SJ joints
joint: SJ 50/30
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
material
stainless steel
Joint
joint: SJ50/30
General description
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.
Choose from one of the most recent versions:
Certificates of Analysis (COA)