Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z153168

Cochranes molecular models

minit, lattices, single kit with booklet

Synonym(s):

chemistry model sets, molecular model sets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104506
NACRES:
NB.17

description

Scale: 2 cm = 0.1 nanometer (1 Å), Minit 3 cm = 0.1 nanometer (1 Å), Orbit

feature

no. of atoms 270
no. of bonds 210

manufacturer/tradename

Cochranes 0074

Looking for similar products? Visit Product Comparison Guide

General description

An extensive model system offering 15 atomic bonding configurations in 12 colors of plastic atoms. The carbon atom is represented in nine configurations: linear, trigonal (in five angle variations), tetrahedral, octahedral, trigonal-bipyramidal. Models come in two sizes: Orbit and Minit. Spare atom centers and bonding tubes are available for all model kits.

Please see Technical Information Bulletin AL-183 for a listing of these items, available on our website at sigma-aldrich.com
Use the 8 and 12 coordinate atom centres within this set to build models of diamond, graphite, sodium chloride, 8 and 12 coordinate metals, caesium chloride, fluorite, zinc blende, wurzite, rutile and ice. Includes 5 instruction/activity booklets.

Scale: 2 cm = 0.1 nanometer (1A), Minit

ATOMS:
  • 2-prong linear: Hydrogen, white (25)
  • 3-prong 120°: Carbon, black (30), Oxygen, red (30)
  • 4-prong tetrahedral: Carbon, black (35), Oxygen, red (15), Sulfur, yellow (20), Halogen, green (10), Metal, silver (25)
  • 6-prong octahedral: Halogen, green (5), Metal, silver (15)
  • 8-prong cubic: Halogen, green (5), Metal, silver (20)
  • 12-prong: metal, silver (20)

BONDS:
  • Straw, green, 2.5 cm (80)
  • Straw, green, 1.5 cm (40)
  • Straw, white, 10 cm (10)
  • Straw, white, 3.5 cm (20)
  • Straw, mixed set/26 (1)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Use our handy VSEPR chart to find the 3-D geometric VSEPR shapes of molecules and ions and learn about VSEPR theory and shapes.

Use our handy VSEPR chart to find the 3-D geometric VSEPR shapes of molecules and ions and learn about VSEPR theory and shapes.

Use our handy VSEPR chart to find the 3-D geometric VSEPR shapes of molecules and ions and learn about VSEPR theory and shapes.

Use our handy VSEPR chart to find the 3-D geometric VSEPR shapes of molecules and ions and learn about VSEPR theory and shapes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service