Key Documents
Z131261
Sigma-Aldrich
Nalgene® volumetric flask
volume 250 mL, accuracy: 0.24 mL, translucent polypropylene copolymer (PPCO)
Synonym(s):
nalgene, plastic flask, plastic flasks
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
long
translucent polypropylene copolymer (PPCO)
Joint
joint threaded neck20mm
sterility
non-sterile
feature
accuracy class B (ASTM E288 and ISO Standard 384)
closure type Screw cap
packaging
pack of 1
manufacturer/tradename
Nalgene® 4000-0250
volume measuring range
250 mL, accuracy: 0.24 mL
capacity
250 mL
General description
Gravimetrically calibrated TC/TD at 20°C individually, then marked with a permanent color graduation line. Meet Class B accuracy requirements of ASTM E288 and ISO Standard 384. No meniscus to confuse readings. Oval marking spot. Linerless PP screw closure. Leakproof and autoclavable.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)