Key Documents
Z125490
Ace-Thred Thermometer/Tubing Adapter
Angled and offset, joint: ST/NS 24/40, thread I.D. 7.0 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Pricing and availability is not currently available.
label
B
Joint
joint: ST/NS24/40
manufacturer/tradename
Ace Glass 503222
thread I.D.
7.0 mm
Related Categories
General description
Threaded nylon bushing with FETFE® O-ring.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)