Key Documents
Z123358
Sigma-Aldrich
Presslok demountable cell holder
for window diameter 25 mm
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
technique(s)
IR spectroscopy: suitable
General description
Assembly is accomplished by compressing cell plates between barrel assembly and self-locking cover. Highly efficient friction lock makes cell leak-proof. For use with all IR spectrometers. Pathlengths are varied by combinations of different PTFE spacers listed. Requires 25 or 32 mm diam. undrilled crystal windows listed (not included).
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)