Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WHA7195002

Whatman® nitrocellulose filter discs 5 μm pore size

white, hydrophilic, 100 ea, 25 mm diam

Synonym(s):

Whatman filter, Z746169, membrane filters, syringe filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

product name

Whatman® nitrocellulose membrane filters, plain white, pore size 5 μm, diam. 25 mm

material

plain white

sterility

non-sterile

packaging

pack of 100 ea

manufacturer/tradename

Whatman 7195-002
Whatman Article No. 28420761 (US reference)

diam.

25 mm

pore size

5 μm

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Narrow pore size distribution for improved surface capture and analysis.
  • Low levels of extractables to ensure sample integrity.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dori M Landry et al.
Genes & development, 23(23), 2753-2764 (2009-12-03)
Most eukaryotic mRNAs are translated using a cap-dependent mechanism of translation. However, approximately 10% of mammalian mRNAs initiate translation using a cap-independent mechanism that is not well understood. These mRNAs contain an internal ribosome entry site (IRES) located in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service