Documents
WHA420200
Whatman® plastic filter holders
Swin-Lok Holder, diam. 25 mm
Synonym(s):
Whatman filter, Whatman filters, Z742686
About This Item
UNSPSC Code:
41104922
NACRES:
NB.24
material
polypropylene housing
sterility
non-sterile
feature
Swin-Lok Holder
packaging
pkg of 10 ea
manufacturer/tradename
Whatman 420200
Whatman Article No. 28419555 (US reference)
diam.
25 mm
General description
Filter holders designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
Features and Benefits
  • Designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
  • All three holders will accommodate Nucleopore track-etched and cast membranes.
  • Syringe compatible.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva