Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W6551

Sodium bicarbonate-13C

API for Clinical Studies, 98 atom % 13C

Synonym(s):

Sodium hydrogencarbonate-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH13CO3
CAS Number:
Molecular Weight:
85.00
MDL number:
UNSPSC Code:
12142200

grade

cGMP

isotopic purity

98 atom % 13C

mp

>300 °C (lit.)

mass shift

M+1

storage temp.

15-25°C

SMILES string

[Na].O[13C](O)=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1/i1+1;

InChI key

UIIMBOGNXHQVGW-YTBWXGASSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service