Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W487012

Sigma-Aldrich

2-Ethyl-4-methyl-1,3-dithiolane

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12S2
CAS Number:
Molecular Weight:
148.29
FEMA Number:
4870
UNSPSC Code:
12164502
Organoleptic:
garlic; horseradish; onion; sulfurous
grade:
Halal
Kosher
biological source:
botanical
food allergen:
no known allergens

biological source

botanical

Quality Level

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 117

Assay

≥95%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

garlic; horseradish; onion; sulfurous

SMILES string

S1C(SCC1C)CC

InChI

1S/C6H12S2/c1-3-6-7-4-5(2)8-6/h5-6H,3-4H2,1-2H3

InChI key

PYBVIKJOGRMFFA-UHFFFAOYSA-N

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

171.5 °F

Flash Point(C)

77.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service