Skip to Content
MilliporeSigma
All Photos(1)

Documents

W464755

Sigma-Aldrich

Geranium thiazole

≥98%, FG

Synonym(s):

4,5-Dimethyl-2-isobutyl thiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NS
CAS Number:
Molecular Weight:
169.29
FEMA Number:
4647
MDL number:
UNSPSC Code:
12164502
Flavis number:
15.078
NACRES:
NA.21

grade

FG
Halal
Kosher

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

geranium; green; nutty; vegetable; earthy

SMILES string

CC(C)Cc1nc(C)c(C)s1

InChI

1S/C9H15NS/c1-6(2)5-9-10-7(3)8(4)11-9/h6H,5H2,1-4H3

InChI key

NSVPHVLZAKJSGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service