Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W420055

Sigma-Aldrich

L-Piperitone

natural, ≥85%

Synonym(s):

(R)-6-(Isopropyl)-3-methylcyclohex-2-en-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
FEMA Number:
4200
MDL number:
UNSPSC Code:
12164502
Flavis number:
07.255
NACRES:
NA.21

grade

Halal
Kosher
natural

Quality Level

reg. compliance

FDA 21 CFR 117

Assay

≥85%

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cooling; minty; peppermint

SMILES string

O=C1[C@H](CCC(=C1)C)C(C)C

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)6-10(9)11/h6-7,9H,4-5H2,1-3H3/t9-/m1/s1

InChI key

YSTPAHQEHQSRJD-SECBINFHSA-N

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F

Flash Point(C)

103 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service