Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W401211

Sigma-Aldrich

1-Methylbutyl acetate

≥98%, FG

Synonym(s):

2-Pentyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O2
CAS Number:
Molecular Weight:
130.18
FEMA Number:
4012
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.657
NACRES:
NA.21
grade:
FG
Halal
Kosher
food allergen:
no known allergens

grade

FG
Halal
Kosher

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; musty; herbaceous; fruity; vegetable

SMILES string

O(C(CCC)C)C(=O)C

InChI

1S/C7H14O2/c1-4-5-6(2)9-7(3)8/h6H,4-5H2,1-3H3

InChI key

GQKZRWSUJHVIPE-UHFFFAOYSA-N

Related Categories

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 - 73.4 °F

Flash Point(C)

21 - 23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service