Skip to Content
MilliporeSigma
All Photos(1)

Documents

W374800

Sigma-Aldrich

L-Menthyl lactate

≥97%, FG

Synonym(s):

Laevo-menthyl lactate, [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2-hydroxypropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24O3
CAS Number:
Molecular Weight:
228.33
FEMA Number:
3748
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.551
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥97%

optical activity

[α]20/D -81°, c = 5 in ethanol

bp

142 °C/5 mmHg (lit.)

mp

42-47 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

mild; cooling

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)[C@H](C)O

InChI

1S/C13H24O3/c1-8(2)11-6-5-9(3)7-12(11)16-13(15)10(4)14/h8-12,14H,5-7H2,1-4H3/t9-,10?,11+,12-/m1/s1

InChI key

UJNOLBSYLSYIBM-SGUBAKSOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Menthyllactate is a flavoring ingredient with a slight minty odor and earthy taste. It provides a long-lasting cooling effect in the mouth. The ingredient has alsobeen proposed as a cosmetic fragrance ingredient .

Application

L-Menthyllactate is used as a cooling ingredient with a pleasant and long-lastingcooling effect in products like toothpaste mints and chewing gum. (2)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemistry and technology of flavours and fragrances
David R
Chemistry and Technology of Flavors and Fragrances. (2009)
Peng Bai et al.
Nature medicine, 25(8), 1266-1273 (2019-07-10)
The ability to safely control transgene expression with simple synthetic gene switches is critical for effective gene- and cell-based therapies. In the present study, the signaling pathway controlled by human transient receptor potential (TRP) melastatin 8 (hTRPM8), a TRP channel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service