Skip to Content
MilliporeSigma
All Photos(1)

Documents

W368806

Sigma-Aldrich

cis-3-Hexenyl benzoate

≥97%, stabilized, FG

Synonym(s):

(Z)-3-hexen-1-yl benzoate, Hexenyl-cis-3-benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
204.26
FEMA Number:
3688
EC Number:
Council of Europe no.:
11778
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.806
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥97%

contains

α-tocopherol as stabilizer (synthetic)

refractive index

n20/D 1.508 (lit.)

bp

105 °C/1 mmHg (lit.)

density

0.999 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; woody; floral

SMILES string

[H]\C(CC)=C(/[H])CCOC(=O)c1ccccc1

InChI

1S/C13H16O2/c1-2-3-4-8-11-15-13(14)12-9-6-5-7-10-12/h3-7,9-10H,2,8,11H2,1H3/b4-3-

InChI key

BCOXBEHFBZOJJZ-ARJAWSKDSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaoyao Peng et al.
Molecules (Basel, Switzerland), 24(11) (2019-05-31)
Feijoa is an aromatic fruit and the essential oil from feijoa peel could be a valuable by-product in the juicing industry. An initial comparison of the essential oil extraction methods, steam-distillation and hydro-distillation, was conducted. The volatile compounds in the
Yaoyao Peng et al.
Food research international (Ottawa, Ont.), 129, 108873-108873 (2020-02-11)
The phenolic compounds and aroma active compounds in feijoa (Acca sellowiana (O.Berg) Burret) juice from four New Zealand grown cultivars (Apollo, Unique, Opal Star, and Wiki Tu) were investigated. A high total phenolic content (maximum 1.89 mg GAE/mL juice) and significant
Béla P Molnár et al.
Frontiers in physiology, 9, 323-323 (2018-04-19)
Insects use sensitive olfactory systems to detect relevant host volatiles and avoid unsuitable hosts in a complex environmental odor landscape. Insects with short lifespans, such as gall midges (Diptera: Cecidomyiidae), are under strong selection pressure to detect and locate suitable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service