Skip to Content
MilliporeSigma
All Photos(1)

Documents

W360301

Sigma-Aldrich

2-Octen-4-one

mixture of isomers, natural, ≥97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O
CAS Number:
Molecular Weight:
126.20
FEMA Number:
3603
MDL number:
UNSPSC Code:
12164502
Flavis number:
07.082
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥97%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; mushroom; fruity; tropical

SMILES string

CCCCC(=O)\C=C\C

InChI

1S/C8H14O/c1-3-5-7-8(9)6-4-2/h4,6H,3,5,7H2,1-2H3/b6-4+

InChI key

FMDLEUPBHMCPQV-GQCTYLIASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

142.0 °F

Flash Point(C)

61.11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Europe offers an attractive and diverse marketplace for flavor formulation. Selling flavors in Europe means adherence in all regions to the EU requirements. Some elements of the EU regulation, such as the community list, may seem daunting, let’s clarify by walking through the requirements

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service