Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W330604

Sigma-Aldrich

5H-5-Methyl-6,7-dihydrocyclopenta[b]pyrazine

≥97%, FG

Synonym(s):

Nutty pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N2
CAS Number:
Molecular Weight:
134.18
FEMA Number:
3306
EC Number:
Council of Europe no.:
2314
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.037
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.527 (lit.)

bp

96-99 °C/20 mmHg (lit.)

density

1.055 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

hazelnut; musty; nutty; peanut

SMILES string

CC1CCc2nccnc12

InChI

1S/C8H10N2/c1-6-2-3-7-8(6)10-5-4-9-7/h4-6H,2-3H2,1H3

InChI key

YZEFQPIMXZVPKP-UHFFFAOYSA-N

Related Categories

General description

5H-5-Methyl-6,7-dihydrocyclopenta[b]pyrazine is the major flavor compound of palm sugar. It is also found in coffee and cooked meat.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changes in volatile compounds of palm sap (Arenga pinnata) during the heating process for production of palm sugar.
Ho CW, et al.
Food Chemistry, 102(4), 1156-1162 (2007)
Pyrazines in foods.
Maga JA, et al.
Critical Reviews in Food Science and Nutrition, 4(1), 39-115 (1973)
Pyrazines in foods: an update
Maga JA & Katz I.
Critical Reviews in Food Science and Nutrition, 16(1), 1-48 (1982)
Cycloalkapyrazines in coffee aroma
Vitzthum OG & Werkhoff P.
Journal of Agricultural and Food Chemistry, 23(3), 510-516 (1975)
Jonita Stankevičiūtė et al.
Scientific reports, 6, 39129-39129 (2016-12-17)
Pyridinols and pyridinamines are important intermediates with many applications in chemical industry. The pyridine derivatives are in great demand as synthons for pharmaceutical products. Moreover, pyridines are used either as biologically active substances or as building blocks for polymers with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service