Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W323705

Sigma-Aldrich

2,3,5,6-Tetramethylpyrazine

≥98%, FG

Synonym(s):

Chuanxingzine, Ligustrazine, Tetrapyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
FEMA Number:
3237
Beilstein:
113100
EC Number:
Council of Europe no.:
734
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.018
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥98%

bp

190 °C (lit.)

mp

77-80 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

chocolate; coffee; fatty; musty; nutty

SMILES string

Cc1nc(C)c(C)nc1C

InChI

1S/C8H12N2/c1-5-6(2)10-8(4)7(3)9-5/h1-4H3

InChI key

FINHMKGKINIASC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Booklice Liposcelis bostrychophila are efficiently attracted by the combination of 2,3,5,6-tetramethylpyrazine and ultraviolet light.: This study demonstrates that the combination of 2,3,5,6-tetramethylpyrazine and ultraviolet light effectively attracts booklice, suggesting a potential application for pest management in stored product environments (Tanaka et al., 2024).

  • 2,3,5,6-Tetramethylpyrazine protects retinal photoreceptors against endoplasmic reticulum stress by modulating ATF4-mediated inhibition of PRP aggregation.: The research highlights the neuroprotective effects of 2,3,5,6-tetramethylpyrazine, showing its potential in treating retinal diseases by protecting photoreceptors from stress-induced damage (Huang et al., 2021).

  • Tetramethylpyrazine-Inducible Promoter Region from Rhodococcus jostii TMP1.: The study identifies a promoter region in Rhodococcus jostii TMP1 that is inducible by tetramethylpyrazine, which could be utilized in genetic engineering and biotechnology applications (Stanislauskienė et al., 2018).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Xian-Chao Cheng et al.
Bioorganic & medicinal chemistry, 17(8), 3018-3024 (2009-03-31)
A series of novel acylpiperazinyl Ligustrazine derivatives was designed, synthesized, and their protective effects on damaged ECV-304 cells and antiplatelet aggregation activities were evaluated. The results showed that compound E33 displayed most potential protective effects on the ECV-304 cells damaged
Hongfei Chen et al.
European journal of medicinal chemistry, 46(11), 5609-5615 (2011-10-14)
A series of novel ligustrazinyloxy-cinnamic acid derivatives were designed, synthesized and evaluated for their inhibitory effect on adenosine diphosphate (ADP)-induced platelet aggregation in vitro, and also assayed for their protective effect against hydrogen peroxide (H(2)O(2))-induced oxidative damage on ECV-304 cells.
Xian-Chao Cheng et al.
Bioorganic & medicinal chemistry, 15(10), 3315-3320 (2007-03-27)
A series of novel Ligustrazine derivatives was designed, synthesized, and assayed for their protective effects on damaged ECV-304 cells and antiplatelet aggregation activities. The results showed that most Ligustrazine derivatives exhibited lower EC(50) values for protective effects on the ECV-304
Yu-Fang Leng et al.
The American journal of Chinese medicine, 40(6), 1229-1239 (2012-12-12)
The Bennett and Xie (1988) model of chronic constriction injury (CCI) investigated the effects of tetramethylpyrazine (TMP) on neuropathic pain-associated behaviors and neuronal apoptosis in the spinal dorsal horn. Fifty-four male rats were randomly divided into sham (group S), CCI
Penglong Wang et al.
Die Pharmazie, 68(9), 782-789 (2013-10-24)
A new anticancer ligustrazine derivative, 3beta-hydroxyolea-12-en-28-oic acid-3,5,6-trimethylpyrazin-2-methyl ester (T-OA, C38H58O3N2), was previously reported. It was synthesized via conjugating the effective antitumor ingredients of a classic traditional Chinese medicine (TCM) formulation. In the present study, anticancer efficacy of T-OA was evaluated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service