Skip to Content
MilliporeSigma
All Photos(1)

Documents

W309818

Sigma-Aldrich

Valeraldehyde

≥97%, FG

Synonym(s):

Pentanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CHO
CAS Number:
Molecular Weight:
86.13
FEMA Number:
3098
Beilstein:
1616304
EC Number:
Council of Europe no.:
93
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.005
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

Assay

≥97%

autoignition temp.

428 °F

refractive index

n20/D 1.394 (lit.)

bp

102-103 °C (lit.)

mp

−92 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

berry; bread; nutty; fruity

storage temp.

2-8°C

SMILES string

[H]C(=O)CCCC

InChI

1S/C5H10O/c1-2-3-4-5-6/h5H,2-4H2,1H3

InChI key

HGBOYTHUEUWSSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Valeraldehyde is one of the key volatile flavor compounds identified in cooked rice, beef fish meat and uncured ham.

Application


  • Waste to treasure: Electrocatalytic upcycling of n-valeraldehyde to octane by Zn-Co bimetallic oxide with atomic level cation defect.: A groundbreaking approach for converting valeraldehyde, a waste product, into valuable octane, showcasing the potential of electrocatalysis in waste management and resource recovery (Qin M et al., 2024).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A fresh look at meat flavor.
Calkins CR & Hodgen JM.
Meat Science, 77(1), 63-80 (2007)
Determination of aldehydes in fish by high-performance liquid chromatography.
da Cunha Veloso MC, et al.
Journal of Chromatographic Science, 39(5), 173-176 (2001)
A comparison of the volatile fractions from cured and uncured meat.
Cross CK & Ziegler P.
Journal of Food Science, 30(4), 610-614 (1965)
François Fenaille et al.
Rapid communications in mass spectrometry : RCM, 18(1), 67-76 (2003-12-23)
Upon hexanal-modification in the presence of NaCNBH(3), the oxidized B chain of insulin becomes mono- and further dialkylated on both the N-terminal and Lys(29) residues. A pseudo-MS(3) study was performed with a triple-quadrupole mass spectrometer on the different modified lysine-containing
Kimberly J Grossman et al.
The European journal of neuroscience, 27(10), 2676-2685 (2008-05-01)
Odor mixtures can produce several qualitatively different percepts; it is not known at which stage of processing these are determined. We asked if activity within the first stage of olfactory processing, the glomerular layer of the olfactory bulb, predicts odor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service