Skip to Content
MilliporeSigma
All Photos(1)

Documents

W295205

Sigma-Aldrich

3-Propylidenephthalide

mixture of cis and trans isomers, ≥96%, FG

Synonym(s):

3-Propylidene-1(3H)-isobenzofuranone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10O2
CAS Number:
Molecular Weight:
174.20
FEMA Number:
2952
EC Number:
Council of Europe no.:
494
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.005
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥96%

refractive index

n20/D 1.586 (lit.)

bp

170 °C/12 mmHg (lit.)

mp

5 °C (lit.)

density

1.122 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

propylidene phthalide

Organoleptic

spicy; herbaceous; warm

SMILES string

CC\C=C1/OC(=O)c2ccccc12

InChI

1S/C11H10O2/c1-2-5-10-8-6-3-4-7-9(8)11(12)13-10/h3-7H,2H2,1H3/b10-5-

InChI key

NGSZDVVHIGAMOJ-YHYXMXQVSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service